Imprimer
Bulb pipettes, BLAUBRAND®, two marks, Class AS, DAkkS certificate
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DAKKS29723 DAKKS29725 DAKKS29727 DAKKS29732 DAKKS29721 DAKKS29722
BRNDDAKKS29723EA 141 EUR
BRNDDAKKS29723 BRNDDAKKS29725 BRNDDAKKS29727 BRNDDAKKS29732 BRNDDAKKS29721 BRNDDAKKS29722
Bulb pipettes, BLAUBRAND®, two marks, Class AS, DAkkS certificate
Pipettes Pipettes jaugées
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Supplied with DAkkS certificate
Certifications: DIN EN ISO 648; DE-M marking