Imprimer
Muffle furnaces, Thermolyne™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F30420C-33 F30420C-33 F30420C-33-80 F30430CM-33 F48020-33 F6020C-33
BARNF30420C-33EA 0 EUR
BARNF30420C-33 455-0339 455-0343 455-0347 455-0341 455-0342
Muffle furnaces, Thermolyne™
Ovens Furnaces (Ashing and Muffle Ovens)
Thermolyne™ muffle furnaces offer heating solutions for a number of application needs, from general laboratory use to industrial applications. Units are ideal for fast heat up and temperature control up to 1200 °C. The furnaces are used for chemical analysis, materials testing, determination of volatiles in waste water treatment plants, industrial plants, pharmaceutical and cosmetic companies, environmental laboratories, agriculture laboratories and paper manufacturers.
  • Fast heat up times and outstanding energy efficiency
  • Range of capacity from 1,3 to 45 litres
  • Four different digital controllers
Thermolyne™ ashing furnaces with air flow control are ideal for coal and coke ashing and temperature control up to 975 °C.
Informations de livraison: Controllers: A1: single set-point control. B1: single set-point control with 1 ramp to set point and a dwell. C1: with 1 stored programme of 8 segments. D1: with 4 stored programmes (16 segments each) and RS232 communication. All controllers (except A1) have mechanical over temperature protection. All models require 240 V, 50/60 Hz.
Attention: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model shown in the table above.