Imprimer
Antibiotic assay discs
Marques : PRAT DUMAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A004242 A006242 A009242 A012242 A013242
PRATA004242EA 68.8 EUR
PRATA004242 PRATA006242 PRATA009242 PRATA012242 PRATA013242
Antibiotic assay discs
Tests microbiologiques Tests sur les antibiotiques
For determining antibiotic suited to the treatment of an infection and for checking their sensitivity to antibiotics by means of the inhibition zone determination method.
  • Very thick cotton fibre papers, for the absorption of test solutions in antibiotic assays
  • 420 g/m² weight