Imprimer
Dehydrated culture media
Marques : PanReac AppliChem
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A7459.1000KG A6666.1000KG
A7459.1000KGEA 234 EUR
A7459.1000KG A6666.1000KG
Dehydrated culture media
Milieux pour microbiologie Milieux pour microbiologie déshydratés
All media, the basal media as well as the special formulations, are prepared only with raw materials of the highest quality and with utmost care. The single components are dried according to the formulation and used anhydrously, to obtain a fine powder after pulverizing. This enables the user to dissolve the powder easily.
  • Media should be stored in a dry place and tightly closed