Imprimer
Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
Fournisseur: Kimberly-Clark
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Kimtech™ Kimwipes
KIMB34155EA 490 EUR
KIMB34155 115-2155 115-0285
Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
Lingettes Lingettes et essuie-mains à usage unique Lingettes en papier pour tâches délicates
These single ply, light-duty wipes can handle a variety of delicate tasks and easily wipe up liquid and dust.
  • Anti-stat dispensing reduces lint and electrostatic discharge and controls usage
  • Portability reduces waste
  • One-at-a-time dispensing (pop-up dispenser)