Imprimer
Cell-free protein synthesis, rapid translation system (RTS) linear template kits
Marques : biotechrabbit
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BR1400801 BR1401201 BR1402401 BR1402201
733-2334EA 457 EUR
733-2334 733-2345 BTRABR1402401 BTRABR1402201
Cell-free protein synthesis, rapid translation system (RTS) linear template kits
Protein Expression Protein Expression Systems
Expression kits for PCR-based generation of linear templates.
RTS Linear Template Kit Plus: For accelerated screening of protein expression templates and conditions
Fast generation of multiple constructs
Efficient in vitro expression (E. coli and insect)
Primers for affinity tags (Strep-tag II/His6-tag)
Signal peptide sequence primers for expression in insects
RTS Linear Template Fab Kit: Template generation Kit for Disulfide E. coli and Insect Kits; PCR based template generation within 1 day for disulfide protein expression
Rapid linear template generation for functional testing of antibody fragments (Fab, scFv) and disulfide bonded proteins
Introduction of an affinity tag for protein purification
Introduction of a signal peptide sequence for translocation into microsomes
Compatible with both Disulfide E. coli and Disulfide Insect Kits
RTS 100 E. coli LinTempGenSet, His-tag: Generate linear templates for rapid screening and optimisation of 6×His-tagged protein expression
Fast template generation without cloning
Contains all regulatory elements in optimal spacing
Create templates with N- or C-terminal 6×His-tag
Primary PCR product can also be cloned into pIVEX vectors
For use with RTS 100 E. coli HY Kit
RTS Wheat Germ LinTempGen Set, His6-tag: Generate linear templates for rapid screening and optimization of 6×His-tagged protein expression
Rapidly create linear expression constructs
Optimised spacing of eukaryotic regulatory elements
Save time; no laborious cloning
Take advantage of flexible positioning of 6×His-tag