Imprimer
Endothelial cell culture medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-22010 C-22011 C-22110 C-22111 C-22210 C-22215 C-22211 C-22216
C-22010EA 190 EUR
C-22010 C-22011 C-22110 C-22111 C-22210 C-22215 C-22211 C-22216
Endothelial cell culture medium
Milieux de culture cellulaire
PromoCell Endothelial Cell Culture Media are low-serum media optimised for the cultivation of endothelial cells from large blood vessels. The media are optimised for primary human cells but can also be used for bovine, porcine, murine, caprine, rabbit, elephant, dog and rat endothelial cells from large vessels as well as for various endothelial cell lines.
Endothelial Medium 2 lacks endothelial cell growth supplement (ECGS, bovine hypothalamic extract) but contains insulin-like growth factor (Long R3 IGF) and vascular endothelial growth factor (VEGF).
Informations de livraison: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. Medium Kit consists of 1× 500 ml bottle basal medium and 1× SupplementPack (individual vials with pre-measured supplements).