Imprimer
Myocyte growth medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-22070 C-22270 C-22275 C-22170
C-22070EA 265 EUR
C-22070 C-22270 C-22275 C-22170
Myocyte growth medium
Milieux de culture cellulaire
PromoCell Myocyte Growth Medium is a low serum medium supplemented with well chosen hormones and growth factors. It was developed for the optimal expansion and maintenance of primary cardiac myocytes in culture.
The medium is optimised for primary human cells but can also be used for primary porcine cardiac myocytes.
Informations de livraison: PromoCell Myocyte Growth Medium 3 is available as Medium (ready to use) or as Medium Kit. The Medium (ready to use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.