Imprimer
Chondrocyte culture medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-27101 C-27111 C-27115
C-27101EA 332 EUR
C-27101 C-27111 C-27115
Chondrocyte culture medium
Milieux de culture cellulaire
PromoCell Chondrocyte Culture Medium is designed for the optimal cultivation of human chondrocytes. The foetal calf serum in the medium is strictly pretested for optimal performance. The medium is optimised for primary human cells but can also be used for primary porcine and rat chondrocytes.
Informations de livraison: PromoCell Chondrocyte Culture Medium 3 is available as Medium (ready to use) or as Medium Kit. The Medium (ready to use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.