Imprimer
Mesenchymal stem cell differentiation medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-28015 C-28013 C-28012 C-28014 C-28016
C-28015EA 451 EUR
C-28015 C-28013 C-28012 C-28014 C-28016
Mesenchymal stem cell differentiation medium
Milieux de culture cellulaire
PromoCell Mesenchymal Stem Cell Differentiation Media efficiently induce differentiation of MSC into adipogenic, chondrogenic, osteogenic or neurogenic lineages, respectively.