Imprimer
Osteoblast growth medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-27001 C-27010
C-27001EA 335 EUR
C-27001 C-27010
Osteoblast growth medium
Milieux de culture cellulaire
PromoCell Osteoblast Growth Medium was developed to establish and maintain human osteoblast cultures. The foetal calf serum in the medium is strictly pretested for optimal performance.
The media are optimised for primary human cells but can also be used for bovine, murine and rat osteoblasts, as well as osteoblast cell lines.
Informations de livraison: The medium (ready-to-use) consists of 1× 500 ml bottle basal medium and 1× vial of SupplementMix. Basal medium also available separately.