Imprimer
Preadipocyte culture Medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-27410 C-27417 C-27415 C-27411
C-27410EA 249 EUR
C-27410 C-27417 C-27415 C-27411
Preadipocyte culture Medium
Milieux de culture cellulaire
PromoCell Preadipocyte Culture Medium is a low-serum medium optimised for the expansion of human preadipocytes.
The media are optimised for primary human cells but can also be used for murine preadipocytes.
Informations de livraison: PromoCell Preadipocyte Culture Medium is available as Medium (ready to use) or as Medium Kit. The Medium (ready to use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.