Imprimer
Skeletal muscle cell culture medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-23265 C-23060 C-23160 C-23260
C-23265EA 218 EUR
C-23265 C-23060 C-23160 C-23260
Skeletal muscle cell culture medium
Milieux de culture cellulaire
PromoCell Skeletal Muscle Cell Culture Medium is a low-serum medium designed for the expansion of human skeletal muscle cells.
The media are optimised for primary human cells but can also be used for rat, murine and rabbit skeletal muscle cells.
Informations de livraison: Growth medium consists of 1× 500 ml bottle of basal medium (ready-to-use) and 1× vial of SupplementMix. Kit consists of 1× 500 ml bottle of basal medium and 1× SupplementPack (individual vials with pre-measured supplements).