Imprimer
Laboratory fume hoods, benchtop, Protector® XL
Fournisseur: LABCONCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
111820021 111610020 111510020
LBCP111820021EA 0 EUR
LBCP111820021 LBCP111610020 LBCP111510020
Laboratory fume hoods, benchtop, Protector® XL
Hottes et coffrets Hottes chimiques
The Protector® XL benchtop laboratory hoods offer superior design flexibility. Their panel liners are supplied in three depths to meet individual installation and application requirements. These are suitable for general chemistry applications.
  • By-pass airflow design
  • Glacier white powder-coated steel exterior
  • Chemical-resistant molded composite panel liner and pre-set baffle(s) with flame spread less than 25 per ASTM E-84
  • Tempered safety glass vertical-rising sash(es) with powder-coated aluminum frame(s)
  • Curved air foil allows air to sweep the work surface while Clean-Sweep™ openings pull inflow air from under the air foil
  • Removable front and side panels and front and interior access panels for access to plumbing and electrical wiring