Imprimer
Bottles, narrow neck, ungrounded neck, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
215-1178EA 74.1 EUR
SCOT211613679 215-1178 SCOT211614478 215-1179 215-1176 SCOT211611707 215-1177 215-1174 215-1175 215-1418
Bottles, narrow neck, ungrounded neck, DURAN®
Flacons Flacons en verre
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes