Imprimer
Collect Ice (i)Xtreme Safety Ankle Boots, Lace-Up
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6246210/38 6246210/39 6246210/40 6246210/41 6246210/42 6246210/43 6246210/44 6246210/45 6246210/46 6246210/47 6246210/48
114-5885EA 114.93 EUR
114-5885 114-5887 114-5886 114-5882 114-5896 114-5897 114-5898 114-5899 114-5900 114-5901 114-5902
Collect Ice (i)Xtreme Safety Ankle Boots, Lace-Up
Chaussures et bottes
Non metallic full grain, leather ankle boot with a strong water resistant upper construction and an innovative PU/nitrile outsole for extrem performances.
  • (i)NTELLIGENT COLD INSULATION: For a strong cold insulation
  • (i)NTEGRATED ANKLE PROTECTION: For additional ankle protection
  • (i)NTELLIGENT WEAR CONTROL: Helps users to detect when the outsole is used
  • Bi-density insole for improved comfort
  • Non metallic anti-puncture midsole
  • Smartium non metallic toecap made from injected polymer
Colour: Brown
EN ISO 20345:2011; S3, SRC, HI, HRO, CI, AN.