Imprimer
Peristaltic pumps, PLP series, behrotest®
Fournisseur: Behr
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BEHRB00210220EA 936 EUR
BEHRB00210220 BEHRB00210218 BEHRB00210217
Peristaltic pumps, PLP series, behrotest®
Pompe Pompes péristaltiques
Models PLP 66, PLP 330 and PLP 1000. These peristaltic pumps include a pump rotor, two compression and four guide rollers.
  • Ready for immediate use
  • Adjustable flow rate
  • No tools requires to change pump tubing
The behrotest peristaltic pumps PLP are suitable for the discontinued transport of watery solutions / non aggressive applications. They find their application in the laboratory, the food industry as well as in the analytical, medicinal or biological field. The RPM (rotation per minute) can be adjusted manually and the flow direction can be amended. These PLP pumps have angle connectors to which tubing can be attached either with or without additional brackets, depending on the tube size.