Imprimer
Chemical resistant gloves, Lapren® 706
Fournisseur: KCL by Honeywell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
706.10 706.7 706.8 706.9
KCHL706.10EA 713 EUR
KCHL706.10 KCHL706.7 KCHL706.8 KCHL706.9
Chemical resistant gloves, Lapren® 706
Gants
These latex gloves incorporate a high quantity of polychloroprene to improve their chemical resistance. A mixture of raw materials ensure a high elasticity as well as good tensile strength and tear resistance.
  • With cotton flock lining, elasticated cuff and anatomical design
  • Excellent sensitivity
  • Good grip when handling wet parts
  • High flexibility at low temperatures
  • High elasticity, good wearing comfort
Ideal for laboratory work, cleaning, manufacturing of small parts, precision engineering and in the electronics industry. The RAL Food approval makes this glove suitable for food handling and processing.
Certifications: EN ISO 374-1:2016 Type A (KLMPST), EN 388:2016 (1020X); Cat. III; AQL < 0,65
Food contact approved