Imprimer
Cryogenic vials, glass
Fournisseur: Hycultec
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B-9001-0 B-9011-0-BLA B-9011-0-W B-9011-0-Y B-9011-0-G B-9011-0-R B-9011-0-B B-9011-0L-W B-9011-0L-Y B-9011-0L-R B-9011-PP-BLA B-9011-PP-W B-9011-PP-Y B-9011-PP-G B-9011-PP-R B-9011-PP-B B-9011-PPL-W B-9011-PPL-Y B-9011-PPL-R B-9011-P-BLA B-9011-P-W B-9011-P-Y B-9011-P-G B-9011-P-R B-9011-P-B B-9011-PL-W B-9011-PL-Y B-9011-PL-R
SANBB-9001-0EA 0 EUR
SANBB-9001-0 SANBB-9011-0-BLA SANBB-9011-0-W SANBB-9011-0-Y SANBB-9011-0-G SANBB-9011-0-R SANBB-9011-0-B SANBB-9011-0L-W SANBB-9011-0L-Y SANBB-9011-0L-R SANBB-9011-PP-BLA SANBB-9011-PP-W SANBB-9011-PP-Y SANBB-9011-PP-G SANBB-9011-PP-R SANBB-9011-PP-B SANBB-9011-PPL-W SANBB-9011-PPL-Y SANBB-9011-PPL-R SANBB-9011-P-BLA SANBB-9011-P-W SANBB-9011-P-Y SANBB-9011-P-G SANBB-9011-P-R SANBB-9011-P-B SANBB-9011-PL-W SANBB-9011-PL-Y SANBB-9011-PL-R
Cryogenic vials, glass
Fioles
Glass vials, with screw cap. These vials are suitable for −20 °C deepfreezing. Screw caps with silicone stoppers make sure that the reagents are stored tight and secure.
  • Available with aluminium / coloured plastic / perforated plastic screw caps