Imprimer
Corning® Lymphocyte Separation Medium, Corning
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
25-072-CV 25-072-CI
25-072-CVEA 155 EUR
25-072-CV MDTC25-072-CI
Corning® Lymphocyte Separation Medium, Corning
Cell Separation Media
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.