ImmunoReagents’ Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes color in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Informations de commande: Products can by customized based on specific assay requirements: contact your VWR representative for more information.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat