Imprimer

Anti-IgG Chicken Polyclonal Antibody (DyLight® 594)

Fournisseur: Immunoreagents
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

IMMRIR1054EA 276 EUR
IMMRIR1054
Anti-IgG Chicken Polyclonal Antibody (DyLight® 594)
Anticorps
The ImmunoReagents' DyLight® 594 conjugates generate red fluorescence and shows wide excitation wavelength separation from green fluorophores for multi-color applications with reduced bleedthrough. The physical characteristics of DyLight® 594 are comparable to other 594 dyes. DyLight® 594 is available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 593 nm / 618 nm Emission Color = Red (Similar Dyes: Alexa Fluor 594, Texas Red)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Informations de commande: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 594
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR