Imprimer

Anti-HDAC7 Mouse Monoclonal Antibody [clone: 9G369]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIH1827-56CEA 601 EUR
USBIH1827-56C
Anti-HDAC7 Mouse Monoclonal Antibody [clone: 9G369]
Anticorps
Type: Primary
Antigen: HDAC7 (histone deacetylase 7)
Clonality: monoclonal
Clone: 9G369
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR