Imprimer

Anti-COL1 Mouse Monoclonal Antibody [clone: 6F204]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIC7510-15DEA 869 EUR
USBIC7510-15D
Anti-COL1 Mouse Monoclonal Antibody [clone: 6F204]
Anticorps
Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: Monoclonal
Clone: 6F204
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Dog, Goat, Human, Pig, Sheep
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR