Imprimer

Anti-SELL Mouse Monoclonal Antibody [clone: FMC46]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIC2415-15MEA 852 EUR
USBIC2415-15M
Anti-SELL Mouse Monoclonal Antibody [clone: FMC46]
Anticorps
Recommended dilution: Flow Cytometry: 1:50-1:100

Type: Primary
Antigen: SELL (selectin L)
Clonality: Monoclonal
Clone: FMC46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Human, Monkey
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR