Imprimer

Anti-UB Mouse Monoclonal Antibody [clone: 13B865]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031436EA 506 EUR
USBI031436
Anti-UB Mouse Monoclonal Antibody [clone: 13B865]
Anticorps
Recommended dilution: Immunochemistry: 1:1000; Immunofluorescence: 1:500

Type: Primary
Antigen: UB (ubiquitin)
Clonality: Monoclonal
Clone: 13B865
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, C. elegans, Chicken, Drosophila, Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR