Imprimer

Anti-Neurofilament M Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031408EA 473 EUR
USBI031408
Anti-Neurofilament M Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: Immunochemistry: 1:5000; Immunofluorescence: 1:500-1:1000; Western Blot: 1:20000

Type: Primary
Antigen: Neurofilament M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Avian, Mammalian, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR