Imprimer

Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]

Fournisseur: Merck Millipore (Oncogene)
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

OP10L-200
OP10L-200EA 679 EUR
OP10L-200
Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]
Anticorps
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 90000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR