Imprimer

Anti-MBP Mouse Monoclonal Antibody [clone: 2H9]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NBP2-22121SS NBP2-22121
NOVUNBP2-22121SSEA 268 EUR
NOVUNBP2-22121SS NOVUNBP2-22121
Anti-MBP Mouse Monoclonal Antibody [clone: 2H9]
Anticorps
Type: Primary
Antigen: MBP (myelin basic protein)
Clonality: monoclonal
Clone: 2H9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR