Imprimer

Anti-VLDLR Mouse Monoclonal Antibody [clone: 6A6]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NBP1-78162 NBP1-78162SS
NOVUNBP1-78162EA 554 EUR
NOVUNBP1-78162 NOVUNBP1-78162SS
Anti-VLDLR Mouse Monoclonal Antibody [clone: 6A6]
Anticorps
The VLDL R Antibody (6A6) from Novus Biologicals is a mouse monoclonal antibody to VLDL R. This antibody reacts with human, mouse, rat, bovine. The VLDL R Antibody (6A6) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: VLDL R
Clonality: Monoclonal
Clone: 6A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR