The Carbonic Anhydrase IX / CA9 Antibody (2D3) from Novus Biologicals is a mouse monoclonal antibody to Carbonic Anhydrase IX / CA9. This antibody reacts with human. The Carbonic Anhydrase IX / CA9 Antibody (2D3) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
CyTOF-ready
Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human