Imprimer

Anti-KDM3A Mouse Monoclonal Antibody [clone: 1E12]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NBP1-51678 NBP1-51678SS
NOVUNBP1-51678EA 554 EUR
NOVUNBP1-51678 NOVUNBP1-51678SS
Anti-KDM3A Mouse Monoclonal Antibody [clone: 1E12]
Anticorps
Mouse Monoclonal JMJD1A Antibody (1E12). Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000

Type: Primary
Antigen: JMJD1A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR