Imprimer

Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-1C6]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NBP1-18965SS NBP1-18965
NOVUNBP1-18965SSEA 255 EUR
NOVUNBP1-18965SS NOVUNBP1-18965
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-1C6]
Anticorps
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-1C6
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR