Imprimer

Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-1D11]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NBP1-18963 NBP1-18963SS
NOVUNBP1-18963EA 554 EUR
NOVUNBP1-18963 NOVUNBP1-18963SS
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-1D11]
Anticorps
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-1D11
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR