Imprimer

Anti-DIO3 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NBP1-05767 NBP1-05767SS
NOVUNBP1-05767EA 638 EUR
NOVUNBP1-05767 NOVUNBP1-05767SS
Anti-DIO3 Rabbit Polyclonal Antibody
Anticorps
The DIO3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DIO3. This antibody reacts with human, mouse, rat, hamster, rabbit. The DIO3 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: DIO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Rabbit
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR