Imprimer

Anti-ENPP1 Goat Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB600-816 NB600-816-25
NOVUNB600-816EA 554 EUR
NOVUNB600-816 NOVUNB600-816-25
Anti-ENPP1 Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: ENPP1 (ectonucleotide pyrophosphatase/phosphodiesterase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR