Imprimer

Anti-NEFL Mouse Monoclonal Antibody [clone: DA2]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB300-132 NB300-132SS
NOVUNB300-132EA 514 EUR
NOVUNB300-132 NOVUNB300-132SS
Anti-NEFL Mouse Monoclonal Antibody [clone: DA2]
Antibodies
Type: Primary
Antigen: NEFL (neurofilament, light polypeptide)
Clonality: monoclonal
Clone: DA2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR