Imprimer

Anti-hnRNP M1-M4 Mouse Monoclonal Antibody [clone: 1D8]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB200-314 NB200-314SS
NOVUNB200-314EA 566 EUR
NOVUNB200-314 NOVUNB200-314SS
Anti-hnRNP M1-M4 Mouse Monoclonal Antibody [clone: 1D8]
Anticorps
Type: Primary
Antigen: hnRNP M1-M4
Clonality: monoclonal
Clone: 1D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR