Imprimer

Anti-Striatin Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-74571 NB110-74571SS
NOVUNB110-74571EA 606 EUR
NOVUNB110-74571 NOVUNB110-74571SS
Anti-Striatin Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Striatin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR