Imprimer
Anti-DISC1 Rabbit Polyclonal Antibody
Fournisseur: Novus Biologicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NB110-40773 NB110-40773SS
NOVUNB110-40773EA 554 EUR
NOVUNB110-40773 NOVUNB110-40773SS
Anti-DISC1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: DISC1 (disrupted in schizophrenia 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat