Imprimer

Anti-BRCA1 Mouse monoclonal antibody unconjugated [clone: RAY]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-598 NB100-598SS
NOVUNB100-598EA 580 EUR
NOVUNB100-598 NOVUNB100-598SS
Anti-BRCA1 Mouse monoclonal antibody unconjugated [clone: RAY]
Anticorps
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: RAY
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR