Imprimer

Anti-CD4 Mouse Monoclonal Antibody [clone: B486A1]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-2181SS NB100-2181
NOVUNB100-2181SSEA 255 EUR
NOVUNB100-2181SS NOVUNB100-2181
Anti-CD4 Mouse Monoclonal Antibody [clone: B486A1]
Anticorps
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: B486A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR