Imprimer

Anti-IgG Goat Polyclonal Antibody (CF™ 543)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

20319 20319-1
20319.EA 359 EUR
20319. 20319-1
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Anticorps
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR