Imprimer

Anti-IgG Chicken Polyclonal Antibody (CF™ 633)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

20224-1 20224
20224-1EA 127 EUR
20224-1 20224.
Anti-IgG Chicken Polyclonal Antibody (CF™ 633)
Anticorps
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Chicken
Isotype: none
Reactivity: Rabbit
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR