Imprimer

Anti-IgG Donkey Polyclonal Antibody (CF™ 350)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

20148 20148-1
20148.EA 359 EUR
20148. 20148-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 350)
Anticorps
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 350 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Sheep
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR