Imprimer

Anti-IgG Goat Polyclonal Antibody (CF™ 488A)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

20020 20020-1
20020.EA 359 EUR
20020. 20020-1
Anti-IgG Goat Polyclonal Antibody (CF™ 488A)
Anticorps
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Chicken
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR