Imprimer

Anti-IgG Goat Polyclonal Antibody (CF™ 488A)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

20011 20011-1
20011.EA 238 EUR
20011. 20011-1
Anti-IgG Goat Polyclonal Antibody (CF™ 488A)
Anticorps
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™488A dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Green fluorescent CF™488A (Ex/Em 490/515 nm) is a superior alternative to FITC (or fluorescein) owing to its exceptional brightness and photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR