Imprimer

Anti-Cell adhesion molecule 4 Rabbit Monoclonal Antibody [clone: EP9563]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB150370-100 AB150370-1ML
ABCAAB150370-100EA 650 EUR
ABCAAB150370-100 ABCAAB150370-1ML
Anti-Cell adhesion molecule 4 Rabbit Monoclonal Antibody [clone: EP9563]
Anticorps
Rabbit monoclonal [EP9563] to Cell adhesion molecule 4.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cell adhesion molecule 4
Clonality: Monoclonal
Clone: EP9563
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR