Imprimer

Anticorps monoclonal anti-ANO1 de lapin [clone : SP31]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB64085-500 AB64085-100 AB64085-1
ABCAAB64085-500EA 769 EUR
ABCAAB64085-500 ABCAAB64085-100 ABCAAB64085-1
Anticorps monoclonal anti-ANO1 de lapin [clone : SP31]
Anticorps
Attention:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type : Primaire
Antigène : ANO1 (anoctamine 1, canal chlorure activé par le calcium)
Clonalité : Monoclonal
Clone : SP31
Conjugaison: Non conjugué
Epitope :
Hôte : Lapin
Isotype: IgG
Réactivité :
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR