Imprimer

Anti-DYNLL1/PIN Rabbit Monoclonal Antibody [clone: EP1660Y]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB51603-100 AB51603-1ML
ABCAAB51603-100EA 738 EUR
ABCAAB51603-100 ABCAAB51603-1ML
Anti-DYNLL1/PIN Rabbit Monoclonal Antibody [clone: EP1660Y]
Anticorps
Rabbit monoclonal [EP1660Y] to DYNLL1/PIN.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: DYNLL1/PIN
Clonality: Monoclonal
Clone: EP1660Y
Conjugation:
Epitope: The epitope for this antibody is on the N-terminus, AA2-14.
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR